Assay Details
Target Gene Details
Entrez Gene ID: | 12269 |
Gene Name: | complement component 4 binding protein |
Gene Aliases: |
AI195242, C4bpa |
Location: |
Chr.1:130635921-130661618 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C4bp | NM_007576.3 | 8 | 1218 | NP_031602.3 |
| AB094486.1 | 8 | 1134 | BAC75872.1 | |
| AK136735.1 | 8 | 1132 | BAE23113.1 | |
| AK149468.1 | 8 | 1225 | BAE28899.1 | |
| AK149477.1 | 8 | 1135 | BAE28904.1 | |
| AK162345.1 | 8 | 1130 | ||
| BC012257.1 | 8 | 1190 | AAH12257.1 | |
| M17122.1 | 8 | 1232 | AAA37312.1 |

Genomic Map