Assay Details
Target Gene Details
Entrez Gene ID: | 12283 |
Gene Name: | calcium binding protein 39 |
Gene Aliases: |
AA408805, AA960512, C78372, MO25, MO25alpha |
Location: |
Chr.1:85793410-85851577 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cab39 | NM_133781.4 | 2 | 488 | NP_598542.3 |
| AK144665.1 | 3 | 321 | BAE25997.1 | |
| AK178270.1 | ||||
| AK206209.1 | ||||
| BC020041.1 | 2 | 150 | AAH20041.1 | |
| CJ162690.1 | ||||
| S51858.1 | 2 | 225 | AAB24801.1 |

Genomic Map