Assay Details
Target Gene Details
Entrez Gene ID: | 636808 |
Gene Name: | contactin associated protein-like 5A |
Gene Aliases: |
Caspr5-1, EG636808 |
Location: |
Chr.1:115684497-116587323 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntnap5a | NM_001077425.1 | NP_001070893.1 |

Genomic Map