Assay Details
Target Gene Details
Entrez Gene ID: | 241175 |
Gene Name: | contactin associated protein-like 5B |
Gene Aliases: |
C230078M14Rik, Caspr5-2 |
Location: |
Chr.1:99772247-100486066 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cntnap5b | NM_172851.2 | NP_766439.2 | ||
| AK082656.1 | BAC38563.1 | |||
| BC145618.1 |
More Information
Set Membership: |
|

Genomic Map