Assay Details
Target Gene Details
Entrez Gene ID: | 26754 |
Gene Name: | COP9 (constitutive photomorphogenic) homolog, subunit 5 (Arabidopsis thaliana) |
Gene Aliases: |
AI303502, CSN5, Jab1, Mov34, Sgn5 |
Location: |
Chr.1:10024600-10038159 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cops5 | NM_001277101.1 | 3 | 546 | NP_001264030.1 |
| NM_013715.2 | 3 | 795 | NP_038743.1 | |
| NR_102282.1 | 3 | 803 | ||
| AF065386.1 | 3 | 548 | AAC17179.1 | |
| AF068223.1 | 3 | 563 | AAF61318.1 | |
| AK002761.1 | 3 | 577 | ||
| AK005728.1 | 3 | 582 | ||
| AK012499.1 | 3 | 582 | BAB28282.1 | |
| AK133073.1 | 3 | 575 | BAE21497.1 | |
| AK146271.1 | 3 | 582 | BAE27030.1 | |
| AK151492.1 | 3 | 581 | BAE30445.1 | |
| BC046753.1 | 3 | 541 | AAH46753.1 | |
| CX240457.1 | ||||
| DV056728.1 | 3 | 602 | ||
| U70736.1 | 3 | 571 | AAD03470.1 |
More Information
Set Membership: |
|

Genomic Map