Assay Details
Target Gene Details
Entrez Gene ID: | 12958 |
Gene Name: | crystallin, beta A2 |
Gene Aliases: |
E130107M19Rik |
Location: |
Chr.1:74889934-74898843 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cryba2 | NM_021541.3 | NP_067516.1 | ||
| AJ272227.1 | CAB75585.1 | |||
| AK021374.1 | BAB32389.1 | |||
| AY160973.1 | AAN78172.1 | |||
| BC052746.1 | AAH52746.1 | |||
| BC056989.1 | AAH56989.1 | |||
| CF740624.1 |

Genomic Map