Assay Details
Target Gene Details
Entrez Gene ID: | 56812 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily B, member 2 |
Gene Aliases: |
2700059H22Rik, Dnajb10, Hsj1, mDj8 |
Location: |
Chr.1:75236403-75245692 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajb2 | NM_001159883.1 | NP_001153355.1 | ||
| NM_001159884.1 | NP_001153356.1 | |||
| NM_001159885.1 | NP_001153357.1 | |||
| NM_020266.2 | NP_064662.2 | |||
| NM_178055.4 | NP_835156.1 | |||
| AB028858.1 | BAA88306.1 | |||
| AK076061.1 | BAC36155.1 | |||
| AK156083.1 | BAE33579.1 | |||
| AK171494.1 | BAE42490.1 | |||
| BC011090.1 | AAH11090.1 | |||
| BY073498.1 | ||||
| BY092620.1 | ||||
| CJ174452.1 |
More Information
Set Membership: |
|

Genomic Map