Assay Details
Target Gene Details
Entrez Gene ID: | 55949 |
Gene Name: | eukaryotic translation elongation factor 1 beta 2 |
Gene Aliases: |
2810017J07Rik, Eef1b |
Location: |
Chr.1:63176831-63180486 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eef1b2 | NM_018796.3 | NP_061266.2 | ||
| AF029844.2 | AAC13264.2 | |||
| AK012756.1 | BAB28447.1 | |||
| AK027911.1 | BAC25661.1 | |||
| AK168191.1 | BAE40151.1 | |||
| BC003899.1 | AAH03899.1 | |||
| BC023139.1 | AAH23139.1 | |||
| BC039635.1 | AAH39635.1 |
More Information
Set Membership: |
|

Genomic Map