Assay Details
Target Gene Details
Entrez Gene ID: | 13518 |
Gene Name: | dystonin |
Gene Aliases: |
2310001O04Rik, A830042E19Rik, AW554249, BP230, BPAG1-n, Bpag, Bpag1, Macf2, ah, athetoid, dt, mKIAA0728, nmf203, nmf339 |
Location: |
Chr.1:33908208-34308662 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dst | NM_001276764.1 | NP_001263693.1 | ||
| NM_133833.3 | NP_598594.2 | |||
| NM_134448.3 | NP_604443.2 | |||
| AF396878.1 | AAK83383.1 | |||
| AF396879.1 | AAK83384.1 | |||
| DQ023311.2 | AAY46942.1 | |||
| DQ463750.1 | ||||
| U22452.1 | AAC52230.1 | |||
| U25158.1 | AAC52231.1 |
More Information
Set Membership: |
|

Genomic Map