Assay Details
Target Gene Details
Entrez Gene ID: | 14060 |
Gene Name: | coagulation factor XIII, beta subunit |
Gene Aliases: |
Cf-13b, Cf13b |
Location: |
Chr.1:139501551-139523756 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F13b | NM_031164.2 | 6 | 901 | NP_112441.2 |
| AK149397.1 | 6 | 888 | BAE28851.1 | |
| BC030166.1 | 6 | 886 | AAH30166.1 | |
| D10071.1 | 6 | 881 | BAA00963.1 |

Genomic Map