Assay Details
Target Gene Details
Entrez Gene ID: | 68187 |
Gene Name: | family with sequence similarity 135, member A |
Gene Aliases: |
4921533L14Rik, AI256732, AI835491 |
Location: |
Chr.1:24010758-24100460 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam135a | NM_026604.4 | NP_080880.4 | ||
| AK019549.1 | BAB31792.2 | |||
| AK034175.1 | BAC28616.1 | |||
| AK044359.1 | BAC31883.1 | |||
| AK087116.1 | ||||
| AK142968.1 | BAE25234.1 | |||
| AK173166.1 | BAD32444.1 | |||
| BC031160.1 | AAH31160.1 | |||
| BC070446.1 | AAH70446.1 | |||
| BC083093.1 | AAH83093.1 |

Genomic Map