Assay Details
Target Gene Details
Entrez Gene ID: | 227377 |
Gene Name: | FERM, RhoGEF and pleckstrin domain protein 2 |
Gene Aliases: |
AI465173, BC009153, D030026M03Rik, Fir, mKIAA0793 |
Location: |
Chr.1:93512104-93621976 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Farp2 | NM_145519.2 | NP_663494.2 | ||
| AK050860.1 | ||||
| AK171713.1 | BAE42626.1 | |||
| AK173028.1 | BAD32306.1 | |||
| AW912106.1 | ||||
| BC009153.1 | AAH09153.1 |

Genomic Map