Assay Details
Target Gene Details
Entrez Gene ID: | 51810 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U |
Gene Aliases: |
AA408410, AI256620, AL024194, AL024437, AW557595, C86794, Hnrpu, SAFA, Sp120, hnRNP U |
Location: |
Chr.1:178321108-178337810 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpu | NM_016805.2 | 10 | 2008 | NP_058085.2 |
| AF073992.1 | 10 | 1945 | AAC26866.1 | |
| AK029944.1 | 10 | 1954 | ||
| AK037517.1 | 10 | 1951 | ||
| AK089058.1 | 10 | 2009 | BAC40725.1 | |
| AK145499.1 | 10 | 2011 | BAE26473.1 | |
| AK145682.1 | 10 | 2011 | ||
| AK159084.1 | 10 | 1977 | BAE34802.1 | |
| AK159614.1 | 10 | 2013 | BAE35231.1 | |
| AK159956.1 | 10 | 1950 | BAE35512.1 | |
| AK162264.1 | BAE36825.1 | |||
| AK165844.1 | 10 | 2008 | BAE38409.1 | |
| AK167200.1 | 10 | 2011 | ||
| AK168657.1 | 10 | 1950 | BAE40513.1 | |
| AK207780.1 | ||||
| BC018353.1 | 10 | 1853 | AAH18353.1 |

Genomic Map