Assay Details
Target Gene Details
Entrez Gene ID: | 51810 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U |
Gene Aliases: |
AA408410, AI256620, AL024194, AL024437, AW557595, C86794, Hnrpu, SAFA, Sp120, hnRNP U |
Location: |
Chr.1:178321108-178337810 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hnrnpu | NM_016805.2 | NP_058085.2 | ||
AF073992.1 | AAC26866.1 | |||
AK029944.1 | ||||
AK037517.1 | ||||
AK089058.1 | BAC40725.1 | |||
AK145499.1 | BAE26473.1 | |||
AK145682.1 | ||||
AK159084.1 | BAE34802.1 | |||
AK159614.1 | BAE35231.1 | |||
AK159956.1 | BAE35512.1 | |||
AK162264.1 | BAE36825.1 | |||
AK165844.1 | BAE38409.1 | |||
AK167200.1 | ||||
AK168657.1 | BAE40513.1 | |||
BC018353.1 | AAH18353.1 |
