Assay Details
Target Gene Details
Entrez Gene ID: | 15528 |
Gene Name: | heat shock protein 1 (chaperonin 10) |
Gene Aliases: |
10kDa, Hsp10, mt-cpn10 |
Location: |
Chr.1:55088148-55091317 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hspe1 | NM_008303.4 | NP_032329.1 | ||
| AK088121.1 | BAC40159.1 | |||
| AK133876.1 | BAE21905.1 | |||
| AK151935.1 | BAE30811.1 | |||
| AV570511.1 | ||||
| BC024385.1 | AAH24385.1 | |||
| BC099385.1 | AAH99385.1 | |||
| CJ238411.1 | ||||
| U09659.1 | AAA62229.1 |
More Information
Set Membership: |
|

Genomic Map