Assay Details
Target Gene Details
Entrez Gene ID: | 70676 |
Gene Name: | GULP, engulfment adaptor PTB domain containing 1 |
Gene Aliases: |
3110030A04Rik, 5730529O06Rik, CED-6, Ced6, GULP, Gulp-2 |
Location: |
Chr.1:44551496-44796838 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gulp1 | NM_028450.3 | NP_082726.2 | ||
| AK014093.1 | BAB29151.2 | |||
| AK017798.1 | BAB30939.1 | |||
| AK028490.1 | ||||
| AK036670.1 | ||||
| BC032154.1 | AAH32154.1 | |||
| BY713087.1 | ||||
| DQ786776.1 |
More Information
Set Membership: |
|

Genomic Map