Assay Details
Target Gene Details
Entrez Gene ID: | 15926 |
Gene Name: | isocitrate dehydrogenase 1 (NADP+), soluble |
Gene Aliases: |
AI314845, AI788952, E030024J03Rik, Id-1, Idh-1, Idpc |
Location: |
Chr.1:65158616-65186479 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Idh1 | NM_001111320.1 | NP_001104790.1 | ||
| NM_010497.3 | NP_034627.3 | |||
| AF020039.1 | AAD02919.1 | |||
| AK087063.1 | BAC39792.1 | |||
| AK149019.1 | BAE28720.1 | |||
| AK151212.1 | BAE30207.1 | |||
| AK159173.1 | BAE34873.1 | |||
| AK160896.1 | BAE36075.1 | |||
| AK167158.1 | BAE39299.1 | |||
| AK167583.1 | BAE39644.1 | |||
| AK168832.1 | ||||
| AK216686.1 | ||||
| BC088986.1 | AAH88986.1 | |||
| DQ403075.1 |
More Information
Set Membership: |
|

Genomic Map