Assay Details
Target Gene Details
Entrez Gene ID: | 140559 |
Gene Name: | immunoglobulin superfamily, member 8 |
Gene Aliases: |
AA033172, AU041109, ESTM34, EWI-2, KCT-4, PGRL, R75384 |
Location: |
Chr.1:172312407-172319841 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igsf8 | NM_080419.1 | NP_536344.1 | ||
| AF411055.1 | AAL02217.1 | |||
| AF439263.1 | AAN63626.1 | |||
| AK140254.1 | ||||
| AK146104.1 | BAE26904.1 | |||
| AK146221.1 | BAE26989.1 | |||
| AK158522.1 | BAE34544.1 | |||
| AK168363.1 | BAE40298.1 | |||
| AK186312.1 | ||||
| AK189184.1 | ||||
| AK190325.1 | ||||
| AK190456.1 | ||||
| AK193134.1 | ||||
| AK193634.1 | ||||
| AK193780.1 | ||||
| AK196050.1 | ||||
| AK198924.1 | ||||
| AK200236.1 | ||||
| AK200799.1 | ||||
| AK200990.1 | ||||
| AK201824.1 | ||||
| AK205206.1 | ||||
| AK207017.1 | ||||
| AK209167.1 | ||||
| AK210016.1 | ||||
| AK216251.1 | ||||
| AK216541.1 | ||||
| AK216980.1 | ||||
| BC019587.1 | AAH19587.1 | |||
| BC026464.1 | AAH26464.1 | |||
| BC026633.1 | AAH26633.1 | |||
| BC048387.1 | AAH48387.1 |
More Information
Set Membership: |
|

Genomic Map