Assay Details
Target Gene Details
Entrez Gene ID: | 16182 |
Gene Name: | interleukin 18 receptor 1 |
Gene Aliases: |
Il18ralpha, Il1rrp |
Location: |
Chr.1:40465552-40500854 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il18r1 | NM_001161842.1 | NP_001155314.1 | ||
| NM_001161843.1 | NP_001155315.1 | |||
| NM_008365.2 | 1 | 162 | NP_032391.1 | |
| AK087129.1 | 1 | 199 | ||
| AK089237.1 | BAC40807.1 | |||
| AK170705.1 | BAE41966.1 | |||
| BC023240.1 | AAH23240.1 | |||
| U43673.1 | 1 | 162 | AAC52437.1 |
More Information
Set Membership: |
|

Genomic Map