Assay Details
Target Gene Details
Entrez Gene ID: | 16174 |
Gene Name: | interleukin 18 receptor accessory protein |
Gene Aliases: |
AcPL, IL-18R-beta, IL-18RAcP, IL-18Rbeta, IL-1RAcPL |
Location: |
Chr.1:40515362-40551707 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il18rap | NM_010553.4 | 12 | 1723 | NP_034683.1 |
| AF077347.1 | 12 | 1732 | AAC72197.1 | |
| BC120598.1 | 12 | 1730 | ||
| BC120600.1 | 12 | 1730 |
More Information
Set Membership: |
|

Genomic Map