Assay Details
Target Gene Details
Entrez Gene ID: | 226922 |
Gene Name: | potassium voltage-gated channel, subfamily Q, member 5 |
Gene Aliases: |
7730402H11, 9230107O05Rik, AA589396, D1Mgi1 |
Location: |
Chr.1:21398399-21962650 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnq5 | NM_001160139.1 | 12 | 1944 | NP_001153611.1 |
| NM_023872.3 | 11 | 1887 | NP_076361.1 | |
| AF263836.1 | 11 | 1362 | AAF73447.1 | |
| AK033079.1 | 5 | 445 | BAC28145.1 | |
| AK147264.1 | 12 | 1947 | ||
| AY679158.1 | 11 | 1526 | AAT76442.1 | |
| EU098138.1 |
More Information
Set Membership: |
|

Genomic Map