Assay Details
Target Gene Details
Entrez Gene ID: | 269180 |
Gene Name: | inositol polyphosphate-4-phosphatase, type I |
Gene Aliases: |
107kDa, 9630012D15, D130048C09Rik, R74740 |
Location: |
Chr.1:37299838-37410740 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Inpp4a | NM_001290797.1 | NP_001277726.1 | ||
NM_001290798.1 | NP_001277727.1 | |||
NM_001290799.1 | NP_001277728.1 | |||
NM_030266.4 | NP_084542.2 | |||
NM_172971.3 | NP_766559.2 | |||
AF317838.1 | AAG40778.1 | |||
AK035867.1 | BAC29220.1 | |||
AK149414.1 | BAE28860.1 | |||
BC120541.1 | ||||
BC144748.1 |
