Assay Details
Target Gene Details
Entrez Gene ID: | 269180 |
Gene Name: | inositol polyphosphate-4-phosphatase, type I |
Gene Aliases: |
107kDa, 9630012D15, D130048C09Rik, R74740 |
Location: |
Chr.1:37299838-37410740 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Inpp4a | NM_001290797.1 | 22 | 2808 | NP_001277726.1 |
NM_001290798.1 | 11 | 1537 | NP_001277727.1 | |
NM_001290799.1 | 11 | 1534 | NP_001277728.1 | |
NM_030266.4 | 22 | 2694 | NP_084542.2 | |
NM_172971.3 | 12 | 1570 | NP_766559.2 | |
AF317838.1 | 20 | 2247 | AAG40778.1 | |
AK035867.1 | 11 | 1538 | BAC29220.1 | |
AK149414.1 | 21 | 2601 | BAE28860.1 | |
BC120541.1 | 12 | 1566 | ||
BC144748.1 | 11 | 1530 |
