Assay Details
Target Gene Details
Entrez Gene ID: | 170771 |
Gene Name: | KH domain containing, RNA binding, signal transduction associated 2 |
Gene Aliases: |
6330586C16Rik, Slim1, Slm1 |
Location: |
Chr.1:32172688-32705224 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Khdrbs2 | NM_133235.2 | 5 | 650 | NP_573498.1 |
| AK078160.1 | 5 | 729 | ||
| BC132117.1 | 5 | 640 | ||
| BC132119.1 | 5 | 640 |
More Information
Set Membership: |
|

Genomic Map