Assay Details
Target Gene Details
Entrez Gene ID: | 16331 |
Gene Name: | inositol polyphosphate-5-phosphatase D |
Gene Aliases: |
SHIP, SHIP-1, SHIP1, SIP-145, p150Ship, s-SHIP |
Location: |
Chr.1:87620312-87720510 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Inpp5d | NM_001110192.2 | NP_001103662.1 | ||
NM_001110193.2 | NP_001103663.1 | |||
NM_010566.3 | NP_034696.2 | |||
NR_122078.1 | ||||
AF125996.1 | AAD37118.1 | |||
AF184912.1 | AAF25823.1 | |||
AF184913.1 | AAF25824.1 | |||
AF228679.1 | AAF69143.1 | |||
AK143560.1 | BAE25436.1 | |||
BC108328.1 | AAI08329.1 | |||
BG144006.1 | ||||
U39203.1 | AAB18937.1 | |||
U51742.1 | AAC52606.1 | |||
U52044.1 | AAC53023.1 |
