Assay Details
Target Gene Details
Entrez Gene ID: | 16331 |
Gene Name: | inositol polyphosphate-5-phosphatase D |
Gene Aliases: |
SHIP, SHIP-1, SHIP1, SIP-145, p150Ship, s-SHIP |
Location: |
Chr.1:87620312-87720510 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Inpp5d | NM_001110192.2 | 5 | 770 | NP_001103662.1 |
NM_001110193.2 | 5 | 773 | NP_001103663.1 | |
NM_010566.3 | 5 | 773 | NP_034696.2 | |
NR_122078.1 | 5 | 770 | ||
AF125996.1 | 5 | 686 | AAD37118.1 | |
AF228679.1 | 5 | 684 | AAF69143.1 | |
AK143560.1 | 5 | 710 | BAE25436.1 | |
BC108328.1 | 5 | 646 | AAI08329.1 | |
U39203.1 | 5 | 722 | AAB18937.1 | |
U51742.1 | 5 | 684 | AAC52606.1 | |
U52044.1 | 5 | 706 | AAC53023.1 |
