Assay Details
Target Gene Details
Entrez Gene ID: | 72568 |
Gene Name: | lin-9 homolog (C. elegans) |
Gene Aliases: |
2700022J23Rik, Bara, Lin-9, TGS, TGS1, mLin-9 |
Location: |
Chr.1:180641150-180690687 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lin9 | NM_001103182.2 | 5 | 550 | NP_001096652.2 |
| NR_120498.1 | 5 | 550 | ||
| AF190325.1 | 5 | 366 | AAQ13712.1 | |
| AK042357.1 | 5 | 332 | BAC31235.1 | |
| AK052618.1 | 5 | 348 | BAC35065.1 | |
| AK089989.1 | 5 | 551 |
More Information
Set Membership: |
|

Genomic Map