Assay Details
Target Gene Details
Entrez Gene ID: | 15064 |
Gene Name: | major histocompatibility complex, class I-related |
Gene Aliases: |
H2ls |
Location: |
Chr.1:155127872-155146813 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mr1 | NM_008209.4 | NP_032235.1 | ||
| AF010448.1 | AAD01444.1 | |||
| AF010449.1 | ||||
| AF010450.1 | ||||
| AF010452.1 | ||||
| AK041171.1 | ||||
| AK042119.1 | BAC31174.1 | |||
| AK137171.1 | ||||
| AK144040.1 | ||||
| AK162712.1 | ||||
| BC026137.1 | AAH26137.1 | |||
| U94989.1 | AAB81345.1 |
More Information
Set Membership: |
|

Genomic Map