Assay Details
Target Gene Details
Entrez Gene ID: | 27395 |
Gene Name: | mitochondrial ribosomal protein L15 |
Gene Aliases: |
HSPC145, MRP-L7, Rpml7 |
Location: |
Chr.1:4773198-4785745 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl15 | NM_001177658.1 | NP_001171129.1 | ||
| NM_025300.4 | NP_079576.1 | |||
| NR_033530.1 | ||||
| AK003026.1 | ||||
| AK003367.1 | BAB22741.1 | |||
| AK010474.1 | BAB26967.1 | |||
| AK010604.1 | BAB27054.1 | |||
| AK011775.1 | ||||
| AK017804.1 | BAB30943.1 | |||
| AK017820.1 | ||||
| AK017824.1 | BAB30958.1 | |||
| AK019175.1 | BAB31589.1 | |||
| AK019244.1 | ||||
| AK019249.1 | BAB31626.1 | |||
| AK150880.1 | BAE29929.1 | |||
| AK160660.1 | BAE35948.1 | |||
| BC021304.1 | AAH21304.1 | |||
| BC027233.1 | AAH27233.1 | |||
| BY083200.1 |
More Information
Set Membership: |
|

Genomic Map