Assay Details
Target Gene Details
Entrez Gene ID: | 64659 |
Gene Name: | mitochondrial ribosomal protein S14 |
Gene Aliases: |
1810032L21Rik, AL022847, Rpms14 |
Location: |
Chr.1:160195260-160201186 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrps14 | NM_025474.3 | NP_079750.1 | ||
| AK007673.1 | BAB25177.1 | |||
| AK010242.1 | BAB26793.1 | |||
| AK083352.1 | BAC38881.1 | |||
| BC044057.1 | AAH44057.1 |
More Information
Set Membership: |
|

Genomic Map