Assay Details
Target Gene Details
Entrez Gene ID: | 94227 |
Gene Name: | peptidase inhibitor 15 |
Gene Aliases: |
P24TI, P25TI, PI-15, SugarCrisp |
Location: |
Chr.1:17601901-17630939 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pi15 | NM_053191.2 | NP_444421.2 | ||
| AF329196.1 | AAK16494.1 | |||
| AK030960.1 | ||||
| AK040961.1 | BAC30762.1 | |||
| AK165584.1 | BAE38275.1 | |||
| BC116705.1 |
More Information
Set Membership: |
|

Genomic Map