Assay Details
Target Gene Details
Entrez Gene ID: | 319263 |
Gene Name: | protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1 |
Gene Aliases: |
8430411F12Rik, A030012M09Rik |
Location: |
Chr.1:7088891-7173628 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pcmtd1 | NM_183028.3 | NP_898849.1 | ||
| AK040848.1 | ||||
| AK079468.1 | ||||
| AK134223.1 | ||||
| AK145008.1 | BAE26182.1 | |||
| AK161921.1 | BAE36635.1 | |||
| AK166166.1 | BAE38606.1 | |||
| AK166593.1 | BAE38879.1 | |||
| AK168606.1 | BAE40473.1 | |||
| AK168705.1 | BAE40547.1 | |||
| BC052741.1 | AAH52741.1 | |||
| BC064801.1 | AAH64801.1 | |||
| BC110360.1 | AAI10361.1 | |||
| BC117941.1 | ||||
| BC117942.1 |
More Information
Set Membership: |
|

Genomic Map