Assay Details
Target Gene Details
Entrez Gene ID: | 98432 |
Gene Name: | PH domain and leucine rich repeat protein phosphatase 1 |
Gene Aliases: |
AI836256, Phlpp, Plekhe1, SCOP, mKIAA0606 |
Location: |
Chr.1:106171869-106394245 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Phlpp1 | NM_133821.3 | 2 | 1712 | NP_598582.3 |
| AB093251.1 | 2 | 471 | BAC41435.1 | |
| AK081093.1 | 3 | 776 | ||
| BC059254.1 | 2 | 373 | AAH59254.1 | |
| CN525246.1 | 2 | 178 |
More Information
Set Membership: |
|

Genomic Map