Assay Details
Target Gene Details
Entrez Gene ID: | 19258 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 4 |
Gene Aliases: |
PTPMEG, Ptn4, TEP, TEP/mPTPMEG, hPTP-MEG |
Location: |
Chr.1:119652467-119837542 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn4 | NM_019933.2 | 25 | 2712 | NP_064317.2 |
| AF106702.1 | 24 | 2540 | AAD22773.1 | |
| AK166169.1 | 6 | 649 | ||
| X82226.1 | CAA57703.1 |
More Information
Set Membership: |
|

Genomic Map