Assay Details
Target Gene Details
Entrez Gene ID: | 227099 |
Gene Name: | postmeiotic segregation increased 1 (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.1:53189187-53297026 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pms1 | NM_153556.1 | NP_705784.1 | ||
| AK140689.1 | BAE24444.1 | |||
| BC028939.1 | AAH28939.1 |
More Information
Set Membership: |
|

Genomic Map