Assay Details
Target Gene Details
Entrez Gene ID: | 18711 |
Gene Name: | phosphoinositide kinase, FYVE finger containing |
Gene Aliases: |
5230400C17Rik, Pip5k, Pip5k3, Pipk5k3, PipkIII, p235 |
Location: |
Chr.1:65186639-65278696 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pikfyve | NM_011086.2 | 10 | 1280 | NP_035216.2 |
| AF102777.1 | 10 | 1260 | AAD10191.1 | |
| AK143565.1 | 9 | 1227 | ||
| AK165350.1 | 9 | 1241 | BAE38145.1 | |
| AK173077.1 | 9 | 1182 | BAD32355.1 |
More Information
Set Membership: |
|

Genomic Map