Assay Details
Target Gene Details
Entrez Gene ID: | 18711 |
Gene Name: | phosphoinositide kinase, FYVE finger containing |
Gene Aliases: |
5230400C17Rik, Pip5k, Pip5k3, Pipk5k3, PipkIII, p235 |
Location: |
Chr.1:65186639-65278696 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pikfyve | NM_011086.2 | NP_035216.2 | ||
| AF102777.1 | AAD10191.1 | |||
| AK080527.1 | ||||
| AK139116.1 | BAE23894.1 | |||
| AK143565.1 | ||||
| AK164361.1 | 2 | 1949 | ||
| AK165350.1 | BAE38145.1 | |||
| AK173077.1 | BAD32355.1 | |||
| BY271210.1 |
More Information
Set Membership: |
|

Genomic Map