Assay Details
Target Gene Details
Entrez Gene ID: | 18986 |
Gene Name: | POU domain, class 2, transcription factor 1 |
Gene Aliases: |
2810482H01Rik, NF-A1, Oct-1, Oct1, Otf-1, Otf1 |
Location: |
Chr.1:165865150-166002672 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pou2f1 | NM_011137.3 | NP_035267.2 | ||
| NM_198932.3 | NP_945150.2 | |||
| NM_198933.3 | NP_945151.2 | |||
| NM_198934.3 | NP_945152.2 | |||
| AF508939.1 | AAM34281.1 | |||
| AJ296212.1 | CAC34943.1 | |||
| AJ489474.1 | CAD35743.1 | |||
| AJ489475.1 | CAD35744.1 | |||
| AK028237.1 | BAC25832.1 | |||
| AY177625.1 | AAO45298.1 | |||
| S65461.1 | AAB28234.1 | |||
| X56230.1 | CAA39679.1 | |||
| X68362.1 | CAA48422.1 | |||
| X68363.1 | CAA48423.1 | |||
| X68364.1 | CAA48424.1 | |||
| X70324.1 | CAA49791.1 |

Genomic Map