Assay Details
Target Gene Details
Entrez Gene ID: | 18802 |
Gene Name: | phospholipase C, delta 4 |
Gene Aliases: |
4921507K24Rik |
Location: |
Chr.1:74541972-74565977 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plcd4 | NM_001081456.1 | NP_001074925.1 | ||
| NM_148937.2 | NP_683739.2 | |||
| AK029561.1 | ||||
| AK140231.1 | BAE24292.1 | |||
| AY033991.1 | AAK61537.1 | |||
| BC066156.1 | AAH66156.1 |
Target Gene Details
Entrez Gene ID: | 77264 |
Gene Name: | zinc finger protein 142 |
Gene Aliases: |
9330177B18Rik, BB154236, mKIAA0236 |
Location: |
Chr.1:74565128-74588211 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp142 | NM_029888.2 | 9 | 7325 | NP_084164.1 |
| AK132550.1 | 9 | 7326 | BAE21230.1 |
More Information
Set Membership: |
|

Genomic Map