Assay Details
Target Gene Details
Entrez Gene ID: | 319455 |
Gene Name: | phospholipase D family, member 5 |
Gene Aliases: |
B230365F16Rik |
Location: |
Chr.1:175962301-176280017 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pld5 | NM_001195816.1 | 2 | 398 | NP_001182745.1 |
| NM_176916.4 | 2 | 702 | NP_795890.2 | |
| AK046294.1 | 2 | 665 | ||
| AK082725.1 | 2 | 298 | ||
| AK136851.1 | 2 | 703 | BAE23146.1 | |
| AK136888.1 | 2 | 703 | BAE23156.1 | |
| AK144116.1 | 2 | 691 | BAE25709.1 | |
| BC100428.1 | 2 | 141 | AAI00429.1 | |
| BU609501.1 | 2 | 367 | ||
| CK626039.1 | 2 | 398 |
More Information
Set Membership: |
|

Genomic Map