Assay Details
Target Gene Details
Entrez Gene ID: | 241075 |
Gene Name: | pleckstrin homology domain containing, family M, member 3 |
Gene Aliases: |
9430067K14Rik, A230102O09Rik, AI449570, DAPR, Plekhm1l |
Location: |
Chr.1:64789120-64956824 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhm3 | NM_001039493.1 | 5 | 2109 | NP_001034582.1 |
| AK034959.1 | 5 | 2031 | BAC28895.1 | |
| AK172220.1 | 4 | 1202 | BAE42889.1 |

Genomic Map