Assay Details
Target Gene Details
Entrez Gene ID: | 226525 |
Gene Name: | RAS protein activator like 2 |
Gene Aliases: |
6530401P13, A330066M24Rik |
Location: |
Chr.1:157135183-157412595 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rasal2 | NM_177644.5 | 4 | 1170 | NP_808312.3 |
| AK039584.1 | 4 | 1170 | ||
| AK086812.1 | 4 | 796 | BAC39748.2 | |
| BC120902.2 | 4 | 513 |
More Information
Set Membership: |
|

Genomic Map