Assay Details
Target Gene Details
Entrez Gene ID: | 213464 |
Gene Name: | retinoblastoma binding protein 5 |
Gene Aliases: |
4933411J24Rik, C330016J05 |
Location: |
Chr.1:132477347-132505665 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbbp5 | NM_172517.2 | NP_766105.2 | ||
| AK049247.1 | BAC33635.1 | |||
| AK154782.1 | BAE32825.1 | |||
| AK170520.1 | ||||
| AK171177.1 | BAE42295.1 | |||
| AK179022.1 | ||||
| AK195903.1 | ||||
| BC057632.1 | AAH57632.1 | |||
| BY193756.1 |

Genomic Map