Assay Details
Target Gene Details
Entrez Gene ID: | 213464 |
Gene Name: | retinoblastoma binding protein 5 |
Gene Aliases: |
4933411J24Rik, C330016J05 |
Location: |
Chr.1:132477347-132505665 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbbp5 | NM_172517.2 | 10 | 1181 | NP_766105.2 |
| AK049247.1 | 10 | 1168 | BAC33635.1 | |
| AK154782.1 | 10 | 1182 | BAE32825.1 | |
| AK170520.1 | 10 | 1179 | ||
| AK171177.1 | 10 | 1186 | BAE42295.1 | |
| BC057632.1 | 11 | 1054 | AAH57632.1 | |
| CV562303.1 | 6 | 614 |

Genomic Map