Assay Details
Target Gene Details
Entrez Gene ID: | 381305 |
Gene Name: | RING CCCH (C3H) domains 1 |
Gene Aliases: |
5730557L09Rik, Gm551, N28103, mKIAA2025 |
Location: |
Chr.1:160906411-160974976 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 19 - Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rc3h1 | NM_001024952.2 | NP_001020123.1 | ||
AK139648.1 | BAE24092.1 | |||
AK173335.1 | BAD32613.1 | |||
AY948287.1 | AAY16368.1 | |||
BC138663.1 | ||||
CJ133431.1 |
