Assay Details
Target Gene Details
Entrez Gene ID: | 56210 |
Gene Name: | REV1 homolog (S. cerevisiae) |
Gene Aliases: |
1110027I23Rik, AU022044, Rev1l |
Location: |
Chr.1:38052786-38129919 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rev1 | NM_019570.3 | NP_062516.2 | ||
| AB057418.1 | BAB64933.1 | |||
| AF179302.1 | AAF23323.1 | |||
| AK048838.1 | ||||
| BC058093.1 | AAH58093.1 |
More Information
Set Membership: |
|

Genomic Map