Assay Details
Target Gene Details
Entrez Gene ID: | 58175 |
Gene Name: | regulator of G-protein signaling 20 |
Gene Aliases: |
2900073E09Rik, Rgsz1 |
Location: |
Chr.1:4909573-5070308 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs20 | NM_001177795.1 | NP_001171266.1 | ||
| NM_001290372.1 | NP_001277301.1 | |||
| NM_021374.5 | NP_067349.2 | |||
| AF191552.1 | AAF05756.1 | |||
| AF191554.1 | AAF05757.1 | |||
| AK013773.1 | BAB28987.2 | |||
| AK047734.1 | ||||
| AK078208.1 | BAC37173.1 | |||
| AK089947.1 | ||||
| AK158864.1 | BAE34700.1 | |||
| BC116925.1 | ||||
| BC116929.1 | ||||
| BI730177.1 | ||||
| BI735670.1 | ||||
| BY127242.1 |
More Information
Set Membership: |
|

Genomic Map