Assay Details
Target Gene Details
Entrez Gene ID: | 24012 |
Gene Name: | regulator of G protein signaling 7 |
Gene Aliases: |
- |
Location: |
Chr.1:175059076-175493238 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs7 | NM_001199003.1 | 15 | 1546 | NP_001185932.1 |
| NM_011880.3 | 15 | 1546 | NP_036010.2 | |
| AF011360.1 | 15 | 1391 | AAC99483.1 | |
| BC051133.1 | 15 | 1501 | AAH51133.1 | |
| DQ139285.1 | 2 | 264 | ||
| DQ139286.1 | 2 | 264 | ||
| DQ139287.1 | ||||
| DQ139288.1 | 3 | 324 | ||
| DQ139289.1 | 2 | 264 |

Genomic Map