Assay Details
Target Gene Details
Entrez Gene ID: | 67792 |
Gene Name: | regulator of G-protein signaling 8 |
Gene Aliases: |
6530413N01Rik |
Location: |
Chr.1:153652953-153697668 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rgs8 | NM_026380.3 | 3 | 460 | NP_080656.2 |
| AK044337.1 | 3 | 334 | BAC31874.1 | |
| AK142301.1 | 3 | 329 | BAE25020.1 | |
| BC065056.1 | 3 | 393 | AAH65056.1 | |
| BC094577.1 | 2 | 76 | AAH94577.1 |
More Information
Set Membership: |
|

Genomic Map