Assay Details
Target Gene Details
Entrez Gene ID: | 20725 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade B, member 8 |
Gene Aliases: |
CAP-2, CAP2, NK10, Spi8, ovalbumin |
Location: |
Chr.1:107589925-107610472 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpinb8 | NM_001159748.1 | NP_001153220.1 | ||
| NM_011459.4 | NP_035589.1 | |||
| AK032659.1 | ||||
| AK036811.1 | BAC29588.1 | |||
| AK048230.1 | BAC33278.1 | |||
| AK148413.1 | BAE28539.1 | |||
| BC025121.1 | AAH25121.1 | |||
| U96703.2 | AAB57815.2 |
More Information
Set Membership: |
|

Genomic Map